Please login to your online account to display your discounted pricing

Novus Biologicals™ IL-27 Protein

$2,309.00 - $7,149.00

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1429075

 
novus biologicals canada
NBP14290705MG
0.5mg Each for $7,149.00

NBP14290701

 
novus biologicals canada
NBP14290701MG
0.1mg Each for $2,309.00
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO