missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ IL-13 Protein

$1,539.00 - $3,343.00

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14291501 Novus Biologicals Canada
NBP14291501MG
0.1mg Each for $1,539.00
NBP1429155 Novus Biologicals Canada
NBP14291505MG
0.5mg Each for $3,343.00
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO