Please login to your online account to display your discounted pricing

Novus Biologicals™ IL-1 beta/IL-1F2 Protein

$1,429.00 - $3,145.00

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1429315

 
novus biologicals canada
NBP14293105MG
0.5mg Each for $3,145.00

NBP14293101

 
novus biologicals canada
NBP14293101MG
0.1mg Each for $1,429.00
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO