missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ IL-1 alpha/IL-1F1 Protein

$329.00 - $626.00

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14323105 Novus Biologicals Canada
NBP1432310005MG
0.005mg Each for $329.00
NBP14323103 Novus Biologicals Canada
NBP1432310025MG
0.025mg Each for $626.00
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO