missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ IFN-gamma Protein

$280.88 - $280.88

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14290001 Novus Biologicals Canada
NBP14290001MG
0.1mg Each for $280.88
Specifications Description & Specifications

Specifications

Unlabeled
Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO