Please login to your online account to display your discounted pricing

Novus Biologicals™ IFN-alpha 4 Protein


Manufacturer: novus biologicals canada  NBP1432010005MG

 View more versions of this product

Catalog No. NBP14320105

  • / Each
 

Description & Specifications

Specifications

Quantity 0.005mg
Source Mouse
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name IFN-alpha 4
Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay
Formulation Sterile liquid; 20mM Tris-HCl, 0.15M NaCl, pH 7.5, 1.0% BSA. 0.22μm filtered.