Please login to your online account to display your discounted pricing

Novus Biologicals Human Various Tissue Microarray Slide

$190.10 - $190.10

Products
Catalog Number Mfr. No. Product Type Price Quantity    

NBP230232

 
novus biologicals
NBP230232
Each for $190.10
Description & Specifications

  • Formalin fixed
  • Tissue condition: Normal/Cancer
  • Species: Human
  • Cross reactivity: Human
  • No. of samples: 59
  • No. of patients: 59
  • Core diameter: 2.0mm
  • Section thickness: 4µm

1 Year