missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Histone H4, Sym-Dimethyl Arg3, Peptide, Biotin

$329.00 - $335.73

Specifications

Gene ID (Entrez) 121504
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP19464310 Novus Biologicals Canada
NBP19464310MG
1mg Each for $335.73
NBP1946435 Novus Biologicals Canada
NBP19464305MG
0.5mg Each for $329.00
Specifications Description & Specifications

Specifications

121504
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO