Please login to your online account to display your discounted pricing

Novus Biologicals™ Histone H3, Trimethyl Lys4, Peptide, Biotin

$329.00 - $332.66

Specifications

Gene ID (Entrez) 126961
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1945515

 
novus biologicals canada
NBP19455105MG
0.5mg Each for $329.00

NBP19455110

 
novus biologicals canada
NBP19455110MG
1mg Each for $332.66
Description & Specifications

Specifications

Gene ID (Entrez) 126961
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO