missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Histone H3, Trimethyl Lys36, Peptide

$338.95 - $569.00

Specifications

Gene ID (Entrez) 126961
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP19456810 Novus Biologicals Canada
NBP19456810MG
1mg Each for $569.00
NBP1945685 Novus Biologicals Canada
NBP19456805MG
0.5mg Each for $338.95
Specifications Description & Specifications

Specifications

126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO