missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Histone H2B Peptide (non-modified a.a. 27-47), Biotin

$222.95 - $329.00

Specifications

Gene ID (Entrez) 440689
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP19468410 Novus Biologicals Canada
NBP19468410MG
1mg Each for $329.00
NBP1946845 Novus Biologicals Canada
NBP19468405MG
0.5mg Each for $222.95
Specifications Description & Specifications

Specifications

440689
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO