missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ GSTM2 Protein

$299.66 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) Blocking, Neutralization, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP15085001 Novus Biologicals Canada
NBP15085001MG
0.1mg Each for $299.66
NBP1508505 Novus Biologicals Canada
NBP15085005MG
0.5mg Each for $802.44
Specifications Description & Specifications

Specifications

Unlabeled
Blocking, Neutralization, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO