missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Glyoxalase I Protein

$254.34 - $583.17

Specifications

Conjugate Unlabeled
For Use With (Application) Blocking, Neutralization, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP19887801 Novus Biologicals Canada
NBP19887801MG
0.1mg Each for $254.34
NBP1988785 Novus Biologicals Canada
NBP19887805MG
0.5mg Each for $583.17
Specifications Description & Specifications

Specifications

Unlabeled
Blocking, Neutralization, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO