missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ GATE-16/GABARAPL2 Protein

$385.90 - $1,066.00

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP15096602 Novus Biologicals Canada
NBP150966025MG
0.25mg Each for $1,066.00
NBP15096605 Novus Biologicals Canada
NBP150966005MG
0.05mg Each for $385.90
Specifications Description & Specifications

Specifications

Unlabeled
SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO