missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Flt-3 Ligand Protein

$226.17 - $363.90

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14316903 Novus Biologicals Canada
NBP1431690025MG
0.025mg Each for $363.90
NBP14316905 Novus Biologicals Canada
NBP1431690005MG
0.005mg Each for $226.17
Specifications Description & Specifications

Specifications

Unlabeled
Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO