Please login to your online account to display your discounted pricing

Novus Biologicals™ FGF basic/FGF2 Protein

$164.00 - $164.00

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14315205

 
novus biologicals canada
NBP1431520005MG
0.005mg Each for $164.00
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO