Please login to your online account to display your discounted pricing

Novus Biologicals™ FGF basic/FGF2 Protein


Manufacturer: novus biologicals canada  NBP1431520025MG

 View more versions of this product

Catalog No. NBP14315203

  • / Each
 

Description & Specifications

Specifications

Quantity 0.025mg
Source Mouse
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name FGF basic/FGF2
Conjugate Unlabeled
For Use With (Application) Immunoassay
Formulation Sterile liquid; phosphate buffered saline, pH 7.2, 150mM NaCl, with 1.0% BSA. 0.22μm filtered.