Please login to your online account to display your discounted pricing

Novus Biologicals™ FGF acidic/FGF1 Protein


Manufacturer: novus biologicals canada  NBP1431530005MG

 View more versions of this product

Catalog No. NBP14315305

  • / Each
 

Description & Specifications

Specifications

Quantity 0.005mg
Source Mouse
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name FGF acidic/FGF1
Conjugate Unlabeled
For Use With (Application) Immunoassay