missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ CXCL8/IL-8 Protein

$659.00 - $2,056.00

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14291801 Novus Biologicals Canada
NBP14291801MG
0.1mg Each for $659.00
NBP1429185 Novus Biologicals Canada
NBP14291805MG
0.5mg Each for $2,056.00
Specifications Description & Specifications

Specifications

Unlabeled
Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO