missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ CCL2/MCP1 Protein

$226.17 - $367.12

Specifications

Conjugate Unlabeled
For Use With (Application) Functional Assay, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14318603 Novus Biologicals Canada
NBP1431860025MG
0.025mg Each for $367.12
NBP14318605 Novus Biologicals Canada
NBP1431860005MG
0.005mg Each for $226.17
Specifications Description & Specifications

Specifications

Unlabeled
Functional Assay, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO