Please login to your online account to display your discounted pricing

Novus Biologicals™ beta-NGF Protein

$160.00 - $160.00

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14317705

 
novus biologicals canada
NBP1431770005MG
0.005mg Each for $160.00
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO