Please login to your online account to display your discounted pricing

Novus Biologicals™ beta-NGF Protein


Manufacturer: novus biologicals canada  NBP1431770005MG

 View more versions of this product

Catalog No. NBP14317705

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Conjugate Unlabeled
Quantity 0.005mg
Formulation Phosphate buffered saline, pH 7.2, 150mM NaCl, with 1.0% BSA. 0.22μm filtered.
For Use With (Application) Immunoassay
Common Name beta-NGF
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.