missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ beta Amyloid, pyro Glu3, Peptide

$192.00 - $329.00

Specifications

For Use With (Application) Western Blot, ELISA
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14497505 Novus Biologicals Canada
NBP144975005MG
0.05mg Each for $192.00
NBP14497510 Novus Biologicals Canada
NBP14497510MG
1mg Each for $329.00
Specifications Description & Specifications

Specifications

Western Blot, ELISA
Store at -80°C. Avoid freeze-thaw cycles.
RUO