missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ beta Amyloid Peptide

$329.00 - $1,358.44

Specifications

Gene ID (Entrez) 351
For Use With (Application) Western Blot, ELISA
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1449671 Novus Biologicals Canada
NBP14496710MG
1mg Each for $329.00
NBP14496710 Novus Biologicals Canada
NBP144967100MG
10mg Each for $1,358.44
Specifications Description & Specifications

Specifications

351
Western Blot, ELISA
Store at -80°C. Avoid freeze-thaw cycles.
RUO