Novus Biologicals AbSelect TCS Purification Antibody Purification Kit
Fast and easy to use kit for purification of IgG fractions from hybridoma supernatants
$816.00 - $816.00
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
8620500 Novus Biologicals
8620500
3 Purifications Each for $816.00
Specifications
Description
AbSelect TCS is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from hybridoma supernatants. The method involves the capture of the antibody on the AbSelect TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
  • Purify antibody from 10 to 50mL of hybridoma TCS
  • 100% compatible with lightning-link range of conjugation kits
  • Supplied with all components and buffers
  • Available with full technical support
Description & Specifications
Specifications
0.0