Please login to your online account to display your discounted pricing

Alfa Aesar™ Novobiocin sodium salt, 20mg/mL in distilled water, sterile-filtered


Quantity 25mL

Manufacturer: alfa aesar™  J67381AC

 View more versions of this product

Catalog No. AAJ67381AC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Novobiocin sodium salt
MDL Number MFCD00066541
CAS 1476-53-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RD5425000
Name Note 20mg/mL in distilled water, sterile-filtered
Physical Form Liquid
Storage Note 1 -30° to -10°C
Quantity 25mL

Specifications

Chemical Name or Material Novobiocin sodium salt
CAS 1476-53-5
Name Note 20mg/mL in distilled water, sterile-filtered
MDL Number MFCD00066541
Accessories

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.

-30° to -10°C