missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ ZOOM™ Carrier Ampholytes DFS Item

Small, soluble molecules with both positive and negative charge groups

$288.00 - $304.00

Specifications

For Use With (Application) 2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Includes ZOOM Carrier Ampholytes are supplied as 10mL bottles
Quantity 10mL
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: The ZOOM IPGRunner Mini-Cell includes the ZOOM IPGRunner Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber The ZOOM IPGRunner Combo Kit includes the ZOOM IPGRunner Mini-Cell, electrode wicks, sealing tape, 10 ZOOM IPGRunner Cassettes, and 12 ZOOM Strips, pH 3-10 NL; ZOOM IPGRunner Retrofit Kit for XCell™ SureLock Mini-Cell includes the ZOOM IPGRunner Buffer Core and Lid; ZOOM IPGRunner Cassettes include electrode wicks and sealing tape


Products ${productFamilyLength}
Catalog Number Mfr. No. pH Price Quantity  
Catalog Number Mfr. No. pH Price Quantity  
ZM0022 Invitrogen™
LSZM0022
pH 4-7 Each for $300.00
ZM0023 Invitrogen™
LSZM0023
pH 6-9 Each for $289.00
ZM0024 Invitrogen™
LSZM0024
pH 9-11 Each for $288.00
ZM0021 Invitrogen™
LSZM0021
pH 3-10 Each for $304.00
Specifications

Description

Use Carrier Ampholytes with the ZOOM IPGRunner™ System which consists of three main components: ZOOM IPGRunner Mini-Cell, ZOOM IPGRunner Cassettes and 7cm ZOOM Strips.

  • ZOOM Carrier Ampholytes sort based on their isoelectric points in an electric field, thereby establishing a pH gradient in solution or contributing to the gradient when used with IPG strips
  • Carrier ampholytes help stabilize the pH gradient and current in IPG strips and aid in protein solubility, resulting in reproducible IEF resolution

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
ZOOM Carrier Ampholytes are supplied as 10mL bottles
10mL