Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ XCell SureLock™ Replacement Parts DFS Item

For the XCell SureLock Mini Blot Module

$70.25 - $638.00

Specifications

For Use With (Equipment) XCell SureLock Mini Blot Module
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Product Type Price Quantity    

EI0012

DFS Item
 
invitrogen™
LSEI0012
Buffer Dam Each for $70.25

EI0010

DFS Item
 
invitrogen™
LSEI0010
Lid Each for $326.00

EI0011

DFS Item
 
invitrogen™
LSEI0011
Wedge Each for $196.00

EI0013

DFS Item
 
invitrogen™
LSEI0013
L-Buffer Dam Each for $568.00

EI9014

DFS Item
 
invitrogen™
LSEI9014
Buffer Core Each for $638.00

EI9022

DFS Item
 
invitrogen™
LSEI9022
Platimum wire Each for $89.25

EI9010

DFS Item
 
invitrogen™
LSEI9010
Cassette Opener Each for $83.00

EI9052

DFS Item
 
invitrogen™
LSEI9052
Sponge pad Each for $79.25
Description & Specifications

Specifications

For Use With (Equipment) XCell SureLock Mini Blot Module

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Shipping condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.