Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ WesternBreeze™ Chromogenic Kits DFS Item

Detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots)

$600.00 - $606.00

Specifications

Conjugate AP (Alkaline Phosphatase) Substrate
Includes Blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chromogenic substrate, wash solutions, and incubation trays. Each kit contains complete reagents for 20 mini-blots.
Quantity 1 kit
View More Specs
Includes:  Blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chromogenic substrate, wash solutions, and incubation trays. Each kit contains complete reagents for 20 mini-blots.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Cross Reactivity Price Quantity    

WB7107

DFS Item
 
invitrogen™
LSWB7107
Goat Each for $600.00

WB7105

DFS Item
 
invitrogen™
LSWB7105
Rabbit Each for $606.00

WB7103

DFS Item
 
invitrogen™
LSWB7103
Mouse Each for $602.00
Description & Specifications

Specifications

Conjugate AP (Alkaline Phosphatase) Substrate
Includes Blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chromogenic substrate, wash solutions, and incubation trays. Each kit contains complete reagents for 20 mini-blots.
Quantity 1 kit

Detection is accomplished with a ready-to-use BCIP/NBT substrate for alkaline phosphatase. Non-fading purple precipitates develop at the protein bands on the membrane.

WesternBreeze™ chromogenic offers:

  • Clear background
  • High sensitivity-low picogram levels detectable
  • High specificity
    One simple protocol-no optimization required

Proteins, Expression, Isolation and Analysis, Western Blotting

Shipping Condition: Wet Ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.