Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ WesternBreeze™ Chemiluminescent Kits DFS Item

Detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots)

$746.00 - $748.00

Specifications

Conjugate AP (Alkaline Phosphatase) Substrate
Includes Blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots.
Quantity 1 kit
View More Specs
Includes:  Blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Cross Reactivity Price Quantity    

WB7106

DFS Item
 
invitrogen™
LSWB7106
Rabbit Each for $748.00

WB7104

DFS Item
 
invitrogen™
LSWB7104
Mouse Each for $748.00

WB7108

DFS Item
 
invitrogen™
LSWB7108
Goat Each for $746.00
Description & Specifications

Specifications

Conjugate AP (Alkaline Phosphatase) Substrate
Includes Blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots.
Quantity 1 kit

Detection is accomplished with a ready-to-use CDP-Star™ chemiluminescent substrate for alkaline phosphatase. Protein bands can be captured either by X-ray film or CDP-Star™ compatible imaging system.

WesternBreeze™ Chemiluminescent Kit offers:

  • High specificity, clean background
  • Ultra-sensitivity-femtogram levels detectable
  • Long-lasting signals-up to 5 days
  • Results in less than 3 hours
  • Permanent photographic image

Proteins, Expression, Isolation and Analysis, Western Blotting

Shipping Condition: Wet Ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.