missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Western-Star™ Immunodetection System DFS Item

High sensitivity, high signal intensity, chemiluminescence-based immunodetection system

$570.00 - $580.00

Specifications

Conjugate AP (Alkaline Phosphatase) Substrate
Quantity 30 blots
Sample Type Protein
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Western-SuperStar™ Reagent Set (containing Blocker, Wash Buffer, Assay Buffer, Substrate, Enhancer, Blot-Loc™ bags, Development Folders and pair of Tweezers) and anti-rabbit secondary antibody conjugated to alkaline phosphatase.


Products ${productFamilyLength}
Catalog Number Mfr. No. Cross Reactivity Price Quantity  
Catalog Number Mfr. No. Cross Reactivity Price Quantity  
T1048 Invitrogen™
LST1048
Rabbit Each for $570.00
T1046 Invitrogen™
LST1046
Mouse Each for $580.00
Specifications

Description

Provides speed, flexibility, control of film exposure time, and the ability to use multiple imaging methods.

  • Most complete western blot detection kit available
  • Image immediately or up to 24 hours later
  • Compatible with multiple membrane types, including PVDF, nitrocellulose, and nylon
  • High signal intensity– ideal for X-ray film and CCD-based imaging
  • High sensitivity and high signal to noise–detect femtogram levels of protein
  • Strip, reprobe, and image at least three times–detect multiple proteins
  • Perform phosphoepitope detection
  • Ideal for confirmation of protein knockdown in siRNA-mediated gene silencing experiments

Proteins, Expression, Isolation and Analysis, Western Blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

AP (Alkaline Phosphatase) Substrate
30 blots
Protein