missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)
Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels
Manufacturer: Invitrogen™ LSLC26755
Requires : The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
Specifications
Running Buffers | |
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis | |
5L |
10X | |
Novex Tris-Glycine SDS Running Buffer (10X) | |
Store at 4° to 25°C |
For Research Use Only. Not for use in diagnostic procedures.