missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ TNF-α Swine ELISA Kit DFS Item

Quantifies natural and recombinant swine TNF-α in swine serum, EDTA plasma, buffered solution, or cell culture medium

$538.00 - $998.00

Specifications

Gene ID (Entrez) 397086
Sample Size 25μL
Sample Type Plasma, Serum, Supernatants
Sensitivity <4pg/mL
Target tumor necrosis factor
View More Specs

Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KSC3011 View Documents Invitrogen™
LSKSC3011
96 assays Each for $538.00
KSC3012 View Documents Invitrogen™
LSKSC3012
192 assays Each for $998.00
Description

Description

TNF-α is produced by macrophages and has many immune effects.

Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation & Function, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

397086
Plasma, Serum, Supernatants
tumor necrosis factor
25μL
<4pg/mL

For Research Use Only. Not for use in diagnostic procedures.