Invitrogen™ Novex TNF- Mouse ELISA Kit DFS Item
Designed to detect and quantify the level of mouse TNF-α in serum, plasma, and supernatant using 96-well plates and a microplate reader
Manufacturer:  Invitrogen™ LSKMC3011
 View more versions of this product
Catalog No. KMC3011
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Colorimetric
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
96 assays
Plasma, Serum, Supernatants
2.5 hrs
21926
<3pg/mL
RUO - research use only
50μL
Tumor necrosis factor