Please login to your online account to display your discounted pricing

Invitrogen™ Novex TNF- Monkey ELISA Kit DFS Item

Quantifies natural and recombinant monkey TNF-α in serum, plasma, buffered solution, or cell culture medium

$526.00 - $980.70

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 7124
Precision <2pg/mL
Regulatory Status RUO - research use only
Sample Size 50μL
View More Specs
Includes:  Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

KPC3012

DFS Item
 
invitrogen™
LSKPC3012
192 assays Each for $980.70

KPC3011

DFS Item
 
invitrogen™
LSKPC3011
96 assays Each for $526.00
Description & Specifications

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 7124
Precision <2pg/mL
Regulatory Status RUO - research use only
Sample Size 50μL
Sample Type Plasma, Serum, Supernatants
Target Tumor necrosis factor
Test Time 4 hrs

TNF-α is produced by macrophages and has many immune effects.

Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay

Shipping Condition: Wet ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.