Invitrogen™ Novex™ Thermo Scientific™ Human IL-2 ELISA Kit DFS Item
Designed to assay IL-2 (human interleukin-2) levels in human serum, plasma (with EDTA, heparin, sodium citrate), urine, and cell culture supernatant samples
Manufacturer:  Invitrogen™ LSEH2IL2
 View more versions of this product
Catalog No. EH2IL2
Specifications
Specifications
NP_000577.2, NM_000586.3
3558
1 plate kit
Chromogenic Detection
Anti-Human IL-2 Precoated 96-well Strip Plate, 1 each; Lyophilized Recombinant Human IL-2 Standard, 2 vials; Standard Diluent, 14mL; Biotinylated Antibody Reagent, 8mL; 30X Wash Buffer, 50mL; Streptavidin-HRP Concentrate, 75μL; HRP Dilution Buffer, 14mL; TMB Substrate Solution, 13mL; Stop Solution, 13mL; Adhesive Plate Covers, 6 each
Documents
For Research Use Only. Not for use in diagnostic procedures.