missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex RANTES Human ELISA Kit DFS Item

Designed to quantify protein levels of human RANTES in plasma, supernatant, and other biological fluids

$526.00 - $526.00

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 6352
Regulatory Status RUO - research use only
Target Chemokine (C-C motif) ligand 5
Test Time 2.5 hrs
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, controls, antibody-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KAC1511 Invitrogen™
LSKAC1511
96 assays Each for $526.00
Specifications

Description

Regulated upon Activation Normal T cell Expressed & Secreted (RANTES) is part of the chemokine superfamily.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Colorimetric
RUO - research use only
2.5 hrs
6352
Chemokine (C-C motif) ligand 5