Please login to your online account to display your discounted pricing

Invitrogen™ Novex RANTES Human ELISA Kit DFS Item


Designed to quantify protein levels of human RANTES in plasma, supernatant, and other biological fluids

Manufacturer: invitrogen™  LSKAC1511

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KAC1511

  • / Each
 

Description & Specifications

Specifications

Test Time 2.5 hrs
For Use With (Application) Colorimetric
Sample Size 100μL
Sample Type Plasma, Supernatants
Precision 2pg/mL
Includes Pre-coated plate, standard, controls, antibody-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Quantity 96 assays
Regulatory Status RUO - research use only
Gene ID (Entrez) 6352
Target Chemokine (C-C motif) ligand 5

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.