Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ NuPAGE™ MOPS SDS Running Buffer (20X) DFS Item


Made with high-purity reagents and are strictly quality controlled

Manufacturer: invitrogen™  LSNP000102

 View more versions of this product

The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. NP000102

  • / Each
 

Description & Specifications

Specifications

Buffer Running Buffers
Quantity 5L
Storage Requirements Store at 4° to 25°C
Includes NuPAGE MOPS SDS Running Buffer (20X)
pH pH 7.7
Concentration 20X
Formulation 50mM MOPS, 50mM Tris Base, 0.1% SDS, 1mM EDTA, pH 7.7
For Use With (Application) 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.