missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ NuPAGE™ MES SDS Running Buffer (20X) DFS Item

Made with high-purity reagents and are strictly quality controlled

Manufacturer:  Invitrogen™ LSNP000202

 View more versions of this product

Catalog No. NP000202



Requires : The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.

Specifications

Specifications

Running Buffers
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
pH 7.3
Store at 4° to 25°C.
20X
NuPAGE MES SDS Running Buffer (20X)
5L
Documents

For Research Use Only. Not for use in diagnostic procedures.