missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ NuPAGE™ 12% Bis-Tris Protein Gels: PROMO DFS Item

Promo offer: 5 Boxes

Pre-cast polyacrylamide gels designed to give optimal separation of small- to medium-sized proteins under denaturing conditions.

Manufacturer:  Invitrogen™ LSNP0343BOX5

 View more versions of this product

Catalog No. NP0343BOX5



Description

Description

  • Unlike traditional Tris-Glycine gels, have a neutral pH environment that minimizes protein modifications
  • Use for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial, as well as day-to-day use
  • Each box contains 10 gels
  • Get separation of proteins in as little as 35 minutes
  • Longer shelf life: can be stored for at least 12 months at room temperature
  • Excellent protein integrity: optimized sample preparation process preserves proteins
  • Wide ranges of molecular weight separation with careful selection of the right gel and running buffer
  • Based upon a Bis-Tris-HCl buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0
  • Can be run in as little as 35 minutes when using the NuPAGE MES Running Buffer
  • Provide sharper bands, higher resolution, and higher accuracy of results

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Bis-Tris
15-well
1.0mm
12%
10 to 80 kd (MOPS Buffer), 3.5 to 40 k (MES Buffer)
15μL
Documents

For Research Use Only. Not for use in diagnostic procedures