Invitrogen™ Novex™ Nitrocellulose Pre-Cut Blotting Membranes DFS Item
Ideal for blotting proteins and nucleic acids
$316.00 - $316.00
Specifications
Quantity 20/Pk.
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
LC2000 Invitrogen™
LSLC2000
Nitrocellulose Pre-Cut Blotting Membrane, 0.2μm pore size Each for $316.00
LC2001 Invitrogen™
LSLC2001
Nitrocellulose Pre-Cut Blotting Membrane, 0.45μm pore size Each for $316.00
Specifications
Description
The 0.2μm Pore Size is ideal for the transfer of low molecular weight proteins (<20kDa) and nucleic acids (<300bp).
  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Contain no support fabrics or detergents
  • Are compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, fluorescence, and radiolabeling
  • Provide high-sensitivity with low background
  • Are supplied in a pre-cut, pre-assembled membrane/filter paper sandwich for convenience
  • A protein's properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces
DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
20/Pk.