Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Nitrocellulose/Filter Paper Sandwich DFS Item

Fits 8.6 x 13.5cm E-PAGE™ Gels

$268.00 - $298.00

Specifications

For Use With (Application) E-PAGE™ Gels
Quantity 16/Pk.
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Product Type Price Quantity    

LC2006

DFS Item
 
invitrogen™
LSLC2006
Nitrocellulose/Filter Paper Sandwich, 0.45μm pore size Each for $268.00

LC2009

DFS Item
 
invitrogen™
LSLC2009
Nitrocellulose/Filter Paper Sandwich, 0.2μm pore size Each for $298.00
Description & Specifications

Specifications

For Use With (Application) E-PAGE™ Gels
Quantity 16/Pk.

  • Designed for western transfer of proteins
  • Pre-cut, extra-thick, absorbent Blotting Paper (2.5mm thickness) for E-PAGE
  • Pore Size: 0.45μm
  • Size: 8.5 x 13.5cm

A protein's properties affect its ability to bind to membrane surfaces.

Proteins, Expression, Isolation and Analysis, Western Blotting

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.