Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ MIP-2 Rat ELISA Kit DFS Item


Quantifies natural and recombinant rat MIP-2 in serum, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKRC1021

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KRC1021

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Colorimetric Detection
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
For Use With (Equipment) Microplate Reader
Quantity 96 assays
Sensitivity <1pg/mL
Accession Number NM_053647.1, NP_446099.1
Incubation Time 3.5 hr.
Gene ID (Entrez) 114105
Sample Type Serum, Supernatants
Marker Gene MIP-2
Sample Size 50μL
Target chemokine (C-X-C motif) ligand 2

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.