missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ MIP-2 Rat ELISA Kit DFS Item

Quantifies natural and recombinant rat MIP-2 in serum, buffered solution, or cell culture medium

$526.00 - $980.00

Specifications

Gene ID (Entrez) 114105
Sample Size 50μL
Sample Type Serum, Supernatants
Sensitivity <1pg/mL
Target chemokine (C-X-C motif) ligand 2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KRC1022 Invitrogen™
LSKRC1022
192 assays Each for $980.00
KRC1021 Invitrogen™
LSKRC1021
96 assays Each for $526.00
Specifications

Description

MIP-2 (macrophage inflammatory protein-2) is a member of the CC family of chemokines.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

114105
Serum, Supernatants
chemokine (C-X-C motif) ligand 2
50μL
<1pg/mL