missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ MCP-1 Rat ELISA Kit DFS Item

Quantifies natural and recombinant rat Monocyte Chemoattractant Protein-1 (MCP-1) in serum, plasma, buffered solution, or cell culture medium

$526.00 - $980.00

Specifications

Gene ID (Entrez) 24770
Sample Size 50μL
Sample Type Plasma, Serum, Supernatants
Sensitivity <8pg/mL
Target chemokine (C-C motif) ligand 2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KRC1011 Invitrogen™
LSKRC1011
96 assays Each for $526.00
KRC1012 Invitrogen™
LSKRC1012
192 assays Each for $980.00
Specifications

Description

MCP-1 attracts recruits monocytes, memory T cells, and dendritic cells to sites of tissue injury and infection.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

24770
Plasma, Serum, Supernatants
chemokine (C-C motif) ligand 2
50μL
<8pg/mL