Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ MCP-1 Rat ELISA Kit DFS Item


Quantifies natural and recombinant rat Monocyte Chemoattractant Protein-1 (MCP-1) in serum, plasma, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKRC1012

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KRC1012

  • / Each
 

Description & Specifications

Specifications

Incubation Time 2.5 hr.
Sample Type Plasma, Serum, Supernatants
Sample Size 50μL
For Use With (Equipment) Microplate Reader
Marker Gene MCP-1
Target chemokine (C-C motif) ligand 2
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Gene ID (Entrez) 24770
Quantity 192 assays
For Use With (Application) Colorimetric Detection
Sensitivity <8pg/mL
Accession Number NM_031530.1, NP_113718.1

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.