Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ MCP-1 Mouse ELISA Kit DFS Item

Quantifies natural and recombinant mouse Monocyte Chemoattractant Protein-1 (MCP-1) in serum, buffered solution, or cell culture medium

$526.00 - $980.70

Specifications

Accession Number NM_011333.3, NP_035463.1
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) 20296
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
View More Specs
Includes:  Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

KMC1011

DFS Item
 
invitrogen™
LSKMC1011
96 assays Each for $526.00

KMC1012

DFS Item
 
invitrogen™
LSKMC1012
192 assays Each for $980.70
Description & Specifications

Specifications

Accession Number NM_011333.3, NP_035463.1
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) 20296
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Incubation Time 3.8 hr.
Marker Gene MCP-1
Sample Size 100μL
Sample Type Serum, Supernatants
Sensitivity <9pg/mL
Target chemokine (C-C motif) ligand 2

MCP-1 attracts recruits monocytes, memory T cells, and dendritic cells to sites of tissue injury and infection.

Applications:

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.