Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ MCP-1 Mouse ELISA Kit DFS Item


Quantifies natural and recombinant mouse Monocyte Chemoattractant Protein-1 (MCP-1) in serum, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKMC1012

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KMC1012

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 20296
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Incubation Time 3.8 hr.
For Use With (Equipment) Microplate Reader
Sample Type Serum, Supernatants
Sample Size 100μL
Quantity 192 assays
Sensitivity <9pg/mL
Accession Number NM_011333.3, NP_035463.1
For Use With (Application) Colorimetric Detection
Marker Gene MCP-1
Target chemokine (C-C motif) ligand 2

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.