Invitrogen™ Luminex™ 200 Performance Verification Kit DFS Item
Controls to verify calibration of MagPlex and MicroPlex classification channels and reporter channel
Manufacturer:  Invitrogen™ LSLX200CONK25
Catalog No. LX200CONK25
Includes: Contains 28 strip wells, LX100/200 Performance Verification Kit CD, Luminex Classification Control Microspheres, MagPlex Classification Control Microspheres, Luminex Reporter Control Microspheres, xMAP Fluidics 1 Microspheres, xMAP Fluidics 2 Microspheres.
Description
Description
For use with Automated Maintenance Plate (AMP). Includes Stripwells for the AMP, new fluidic controls for early detection and lot-specific CD for importing target values. Each provides 25 verifications.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
28 strip wells
25 Reactions
Luminex™ 200™ System
SDS
Documents
For Research Use Only. Not for use in diagnostic procedures.